Learn More
Medchemexpress LLC KTX-582 | 2573298-13-0 | 98.7% | 872.93 | 5 MG
SDP
Catalog No. 5000362389
Encompass_Preferred
KTX-582 is a potent IRAK4 degrader that can induce apoptosis in MYD88MT DLBCL and is efficient in inducing in vivo tumor regressions in a lymphoma model. It degrades IRAK4 in whole blood monocytes and lymphocytes with high potency.
  • Potent IRAK4 degrader with DC50 values of 4 nM for IRAK4 and 5 nM for Ikaros
  • Induces apoptosis in MYD88MT DLBCL
  • Efficient in inducing in vivo tumor regressions in a lymphoma model
  • Degrades IRAK4 in whole blood monocytes and lymphocytes with IC50s of less than 0.05 μM
  • Inhibits IRAK4 in human whole blood LPS TNFα with an IC50 of 0.05 to 1 μM
  • Exhibits antiproliferative activity against human OCI-Ly10 cells (IC50: 14 nM)
Catalog No. 50-003-62389 Supplier Medchemexpress LLC Supplier No. HY1482745MG
KTX-582 is a potent IRAK4 degrader that can induce apoptosis in MYD88MT DLBCL and is efficient in inducing in vivo tumor regressions in a lymphoma model. It degrades IRAK4 in whole blood monocytes and lymphocytes with high potency.
  • Potent IRAK4 degrader with DC50 values of 4 nM for IRAK4 and 5 nM for Ikaros
  • Induces apoptosis in MYD88MT DLBCL
  • Efficient in inducing in vivo tumor regressions in a lymphoma model
  • Degrades IRAK4 in whole blood monocytes and lymphocytes with IC50s of less than 0.05 μM
  • Inhibits IRAK4 in human whole blood LPS TNFα with an IC50 of 0.05 to 1 μM
  • Exhibits antiproliferative activity against human OCI-Ly10 cells (IC50: 14 nM)