Learn More
Medchemexpress LLC KTX-582 intermediate-1 | 845508-29-4 | 326.41 g/mol | C16H22O5S | 10 MG
SDP
Catalog No. 5000202895
Encompass_Preferred
KTX-582 intermediate-1 is a synthetic chemical intermediate supplied for research use and intended for use in the synthesis of IRAK4-degrading compounds and antibody-drug conjugate preparation. The material is provided as a powder and requires controlled storage conditions for stability.
  • used as a synthetic intermediate for research and development.
  • intended for laboratory research use only; not for human or clinical applications.
  • powder form with recommended storage conditions to preserve stability.
  • supports synthetic workflows including ADC preparation and targeted degradation studies.
Catalog No. 50-002-02895 Supplier Medchemexpress LLC Supplier No. HY4943610MG
KTX-582 intermediate-1 is a synthetic chemical intermediate supplied for research use and intended for use in the synthesis of IRAK4-degrading compounds and antibody-drug conjugate preparation. The material is provided as a powder and requires controlled storage conditions for stability.
  • used as a synthetic intermediate for research and development.
  • intended for laboratory research use only; not for human or clinical applications.
  • powder form with recommended storage conditions to preserve stability.
  • supports synthetic workflows including ADC preparation and targeted degradation studies.