Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KTX-582 intermediate-2 | 2434850-07-2 | 495.37 | C21H27BrN4O5 | 10 MG
SDP

Catalog No. 5000202109
Encompass_Preferred

KTX-582 intermediate-2 is a synthetic intermediate used in the preparation of IRAK4 degrader molecules and for antibody-drug conjugate (ADC) synthesis. Supplied for research and synthetic applications.

  • Used as a synthetic intermediate for IRAK4 degrader development
  • Molecular weight 495.37
  • Chemical formula C21H27BrN4O5
  • CAS number 2434850-07-2
  • Storage: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-002-02109 Supplier Medchemexpress LLC Supplier No. HY4943210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KTX-582 intermediate-2 is a synthetic intermediate used in the preparation of IRAK4 degrader molecules and for antibody-drug conjugate (ADC) synthesis. Supplied for research and synthetic applications.

  • Used as a synthetic intermediate for IRAK4 degrader development
  • Molecular weight 495.37
  • Chemical formula C21H27BrN4O5
  • CAS number 2434850-07-2
  • Storage: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.