Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KTX-582 intermediate-2 | 2434850-07-2 | 495.37 g/mol | C21H27BrN4O5 | 25 MG
SDP

Catalog No. 5000202896
Encompass_Preferred

KTX-582 intermediate-2 is a synthetic chemical intermediate used in the preparation of KTX-582 and for antibody-drug conjugate (ADC) construction. It is used in medicinal chemistry applications related to IRAK4 degrader development.

  • Intermediate for synthesis and ADC preparation.
  • CAS number 2434850-07-2.
  • Molecular formula C21H27BrN4O5.
  • Molecular weight 495.37 g/mol.
  • Sold in small research quantities; supplier catalog indicates a 25 MG package.

Catalog No. 50-002-02896 Supplier Medchemexpress LLC Supplier No. HY4943225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KTX-582 intermediate-2 is a synthetic chemical intermediate used in the preparation of KTX-582 and for antibody-drug conjugate (ADC) construction. It is used in medicinal chemistry applications related to IRAK4 degrader development.

  • Intermediate for synthesis and ADC preparation.
  • CAS number 2434850-07-2.
  • Molecular formula C21H27BrN4O5.
  • Molecular weight 495.37 g/mol.
  • Sold in small research quantities; supplier catalog indicates a 25 MG package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.