Learn More
Medchemexpress LLC KTX-582 intermediate-2 | 2434850-07-2 | 96.2% | 495.37 g·mol⁻¹ | C21H27BrN4O5 | 5 MG
SDP
Catalog No. 5000202786
Encompass_Preferred
KTX-582 intermediate-2 is a chemical intermediate used in the synthesis of IRAK4 degrader compounds and can be employed for antibody-drug conjugate (ADC) preparation. It is supplied as a solid with reported formula C21H27BrN4O5, molecular weight 495.37 g·mol⁻¹, and CAS number 2434850-07-2. Purity is batch-dependent and is provided on the supplier data sheet.
  • Used as an intermediate in the synthesis of IRAK4 degrader compounds.
  • Suitable for antibody-drug conjugate (ADC) preparation.
  • Supplied in small-scale quantities suitable for research use.
  • Reported molecular formula C21H27BrN4O5 and molecular weight 495.37 g·mol⁻¹.
  • CAS number 2434850-07-2 for unambiguous identification.
Catalog No. 50-002-02786 Supplier Medchemexpress LLC Supplier No. HY494325MG
KTX-582 intermediate-2 is a chemical intermediate used in the synthesis of IRAK4 degrader compounds and can be employed for antibody-drug conjugate (ADC) preparation. It is supplied as a solid with reported formula C21H27BrN4O5, molecular weight 495.37 g·mol⁻¹, and CAS number 2434850-07-2. Purity is batch-dependent and is provided on the supplier data sheet.
  • Used as an intermediate in the synthesis of IRAK4 degrader compounds.
  • Suitable for antibody-drug conjugate (ADC) preparation.
  • Supplied in small-scale quantities suitable for research use.
  • Reported molecular formula C21H27BrN4O5 and molecular weight 495.37 g·mol⁻¹.
  • CAS number 2434850-07-2 for unambiguous identification.