Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KTX-951 | 2573298-36-7 | 99.6% | 850.95 | 1 MG
SDP

Catalog No. 5000359734
Encompass_Preferred

KTX-951 is an orally active PROTAC degrader targeting IRAK4 and IMiD (Ikaros/Aiolos) substrates. It exhibits a Kd of 3.5 nM for IRAK4, with DC50 values of 13 nM, 14 nM, and 13 nM for IRAK4, Ikaros, and Aiolos, respectively. This compound also shows an IC50 of 35 nM for OCl-Ly10 CTG and demonstrates antitumor activity.

  • Orally active PROTAC degrader
  • Targets IRAK4 and IMiD (Ikaros/Aiolos) substrates
  • Kd of 3.5 nM for IRAK4
  • Molecular weight: 850.95
  • Formula: C46H52F2N8O6
  • Appearance: Solid, light yellow to yellow
  • Purity: 99.60%
  • Soluble in DMSO at 50 mg/mL (58.76 mM)
  • Oral bioavailability in rats: 22% at 10 mg/kg
  • Oral bioavailability in dogs: 57% at 10 mg/kg

Catalog No. 50-003-59734 Supplier Medchemexpress LLC Supplier No. HY1482901MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KTX-951 is an orally active PROTAC degrader targeting IRAK4 and IMiD (Ikaros/Aiolos) substrates. It exhibits a Kd of 3.5 nM for IRAK4, with DC50 values of 13 nM, 14 nM, and 13 nM for IRAK4, Ikaros, and Aiolos, respectively. This compound also shows an IC50 of 35 nM for OCl-Ly10 CTG and demonstrates antitumor activity.

  • Orally active PROTAC degrader
  • Targets IRAK4 and IMiD (Ikaros/Aiolos) substrates
  • Kd of 3.5 nM for IRAK4
  • Molecular weight: 850.95
  • Formula: C46H52F2N8O6
  • Appearance: Solid, light yellow to yellow
  • Purity: 99.60%
  • Soluble in DMSO at 50 mg/mL (58.76 mM)
  • Oral bioavailability in rats: 22% at 10 mg/kg
  • Oral bioavailability in dogs: 57% at 10 mg/kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.