Learn More
Medchemexpress LLC KTX-951 | 2573298-36-7 | 99.6% | 850.95 | 1 ML
SDP
Catalog No. 5000360530
Encompass_Preferred
KTX-951 is an orally active PROTAC degrader targeting IRAK4 and IMiD (Ikaros/Aiolos) substrates. It demonstrates potent DC50s against IRAK4, Ikaros, and Aiolos, and exhibits antitumor activity with an IC50 of 35 nM for OCl-Ly10 CTG.
  • Orally active PROTAC degrader
  • Targets IRAK4 and IMiD substrates (Ikaros/Aiolos)
  • Potent DC50s for IRAK4, Ikaros, and Aiolos
  • Exhibits antitumor activity
Catalog No. 50-003-60530 Supplier Medchemexpress LLC Supplier No. HY14829010MM/1ML
KTX-951 is an orally active PROTAC degrader targeting IRAK4 and IMiD (Ikaros/Aiolos) substrates. It demonstrates potent DC50s against IRAK4, Ikaros, and Aiolos, and exhibits antitumor activity with an IC50 of 35 nM for OCl-Ly10 CTG.
  • Orally active PROTAC degrader
  • Targets IRAK4 and IMiD substrates (Ikaros/Aiolos)
  • Potent DC50s for IRAK4, Ikaros, and Aiolos
  • Exhibits antitumor activity