Learn More
Enzo Life Sciences Ku-0063794 (5mg). CAS: 938440-64-3
SDP
Catalog No. 502013429
Encompass_Preferred
Alternative name: rel-5-[2-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-4-(4-morpholinyl)pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol, mTOR Inhibitor IV. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3429 Supplier Enzo Life Sciences Supplier No. ENZCHM1350005
Alternative name: rel-5-[2-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-4-(4-morpholinyl)pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol, mTOR Inhibitor IV. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.