Learn More
Selleck Chemical LLC KU-55933 S1092-200mg
SDP
Catalog No. 503111071
Encompass_Preferred
KU-55933 is a potent and specific ATM inhibitor with IC50/Ki of 12 9 nM/2 2 nM in cell-free assays and is highly selective for ATM as compared to DNA-PK PI3K/PI4K ATR and mTOR KU 55933 (ATM Kinase Inhibitor) inhibits the activation of autophagy initiating kinase ULK1 and results in a significant decrease of autophagy
Catalog No. 50-311-1071 Supplier Selleck Chemical LLC Supplier No. S1092200MG
KU-55933 is a potent and specific ATM inhibitor with IC50/Ki of 12 9 nM/2 2 nM in cell-free assays and is highly selective for ATM as compared to DNA-PK PI3K/PI4K ATR and mTOR KU 55933 (ATM Kinase Inhibitor) inhibits the activation of autophagy initiating kinase ULK1 and results in a significant decrease of autophagy