Learn More
Selleck Chemical LLC KU-55933 (ATM Kinase Inhibitor) 10mM/1mL 587871-26-9
SDP
Supplier:  Selleck Chemical LLC S109210MM/1ML
Encompass_Preferred
KU-55933 (ATM Kinase Inhibitor) is a potent and specific ATM inhibitor with IC50/Ki of 12.9 nM/2.2 nM in cell-free assays, and is highly selective for ATM as compared to DNA-PK, PI3K/PI4K, ATR and mTOR. KU?55933 (ATM Kinase Inhibitor) inhibits the activation of autophagy?initiating kinase ULK1 and results in a significant decrease of autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0964968