Learn More
TARGETMOL CHEMICALS INC KU-57788 25MG
SDP
Catalog No. 502579460
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NU7441 (KU-57788 (NU7441)) is a highly effective and specific DNA-PK inhibitor (IC50 14 nM). purity: 99%
Catalog No. 50-257-9460 Supplier TARGETMOL CHEMICALS INC Supplier No. T627625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NU7441 (KU-57788 (NU7441)) is a highly effective and specific DNA-PK inhibitor (IC50 14 nM). purity: 99%