Learn More
Medchemexpress LLC HY-11006 100mg , KU-57788 NU7441 CAS:503468-95-9 Purity:98%
SDP
Catalog No. 501492851
Encompass_Preferred
Medchemexpress, HY-11006 100mg KU-57788 NU7441 CAS:503468-95-9 Formula:C25H19NO3S IC50: 13 nM (DNA-PK) , 1 μM (BRD4), 3.5 μM (BRDT) Purity:98% KU-57788 is a potent and selective inhibitor of DNA-PK , with an IC 50 of 13 nM, and also increases CRISPR/Cas9 -mediated editing frequencies. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2851 Supplier Medchemexpress LLC Supplier No. HY11006100MG
Medchemexpress, HY-11006 100mg KU-57788 NU7441 CAS:503468-95-9 Formula:C25H19NO3S IC50: 13 nM (DNA-PK) , 1 μM (BRD4), 3.5 μM (BRDT) Purity:98% KU-57788 is a potent and selective inhibitor of DNA-PK , with an IC 50 of 13 nM, and also increases CRISPR/Cas9 -mediated editing frequencies. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.