Learn More
Apexbio Technology LLC KU-60019 925701-49-1 10mg
SDP
Catalog No. 501014294
Encompass_Preferred
KU-60019 is a selective inhibitor targeting ataxia telangiectasia mutated (ATM) kinase with an IC50 value of approximately 6 3 nM Through inhibition of ATM kinase activity KU-60019 influences cellular DNA damage responses and sensitivity to radiation therapy It has been demonstrated to enhance radiosensitivity in human fibroblast cells expressing functional ATM but not in ATM-deficient cells Additionally KU-60019 reduces radiation-induced phosphorylation of AKT at Ser473 in glioma cell lines independently affecting cell migration invasion and proliferation KU-60019 serves as an investigational tool for exploring ATM-dependent signaling pathways and examining therapeutic strategies against cancers involving ATM-regulated DNA repair and cell survival mechanisms
Catalog No. 50-101-4294 Supplier Apexbio Technology LLC Supplier No. A833610
KU-60019 is a selective inhibitor targeting ataxia telangiectasia mutated (ATM) kinase with an IC50 value of approximately 6 3 nM Through inhibition of ATM kinase activity KU-60019 influences cellular DNA damage responses and sensitivity to radiation therapy It has been demonstrated to enhance radiosensitivity in human fibroblast cells expressing functional ATM but not in ATM-deficient cells Additionally KU-60019 reduces radiation-induced phosphorylation of AKT at Ser473 in glioma cell lines independently affecting cell migration invasion and proliferation KU-60019 serves as an investigational tool for exploring ATM-dependent signaling pathways and examining therapeutic strategies against cancers involving ATM-regulated DNA repair and cell survival mechanisms