Learn More
Abcam KU-60019, ATM kinase inhibitor, 50MG
Catalog No. 502705500
Encompass_Preferred
MW 547.7 Da, Purity >98%. Potent, highly selective ATM kinase inhibitor (IC₅₀ = 6.3 nM). Inhibits migration and invasion of glioma cells. Shows similar target selectivity to KU-55933 (ab120637). Shows radiosensitizing effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5500 Supplier Abcam Supplier No. AB14481750MG
MW 547.7 Da, Purity >98%. Potent, highly selective ATM kinase inhibitor (IC₅₀ = 6.3 nM). Inhibits migration and invasion of glioma cells. Shows similar target selectivity to KU-55933 (ab120637). Shows radiosensitizing effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement