Learn More
Selleck Chemical LLC KU-60019 10mg 925701-49-1
SDP
Catalog No. 508689
Encompass_Preferred
KU-60019 is an improved analogue of KU-55933, with IC50 of 6.3 nM for ATM in cell-free assays, 270- and 1600-fold more selective for ATM than DNA-PK and ATR,and is a highly effective radiosensitizer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-868-9 Supplier Selleck Chemical LLC Supplier No. S157010MG
KU-60019 is an improved analogue of KU-55933, with IC50 of 6.3 nM for ATM in cell-free assays, 270- and 1600-fold more selective for ATM than DNA-PK and ATR,and is a highly effective radiosensitizer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.