Learn More
Selleck Chemical LLC KU-60019 10mM/1mL 925701-49-1
SDP
Supplier:  Selleck Chemical LLC S157010MM/1ML
Encompass_Preferred
KU-60019 is an improved analogue of KU-55933, with IC50 of 6.3 nM for ATM in cell-free assays, 270- and 1600-fold more selective for ATM than DNA-PK and ATR,and is a highly effective radiosensitizer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0964969