Learn More
Enzo Life Sciences KU55933 (10mg). CAS: 587871-26-9
SDP
Catalog No. 502013454
Encompass_Preferred
KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer. Alternative name: ATM kinase inhibitor, 2-Morpholino-6-(thianthren-1-yl)-4H-pyran-4-one. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (50 mg/ml) or ethanol (<13 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-3454 Supplier Enzo Life Sciences Supplier No. ENZCHM1500010
KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer. Alternative name: ATM kinase inhibitor, 2-Morpholino-6-(thianthren-1-yl)-4H-pyran-4-one. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (50 mg/ml) or ethanol (<13 mg/ml). Long Term Storage: -20°C.