Learn More
ATCC KU812; Leukemia (CML); Human (Homo sapiens)
Catalog No. 502383065
Encompass
KU812 are basophils that were isolated from the peripheral blood of a 38-year-old, Asian, male patient with chronic myelogenous leukemia (cml). It can be used for immunology and immune system disorder research.
Catalog No. 50-238-3065 Supplier ATCC Supplier No. CRL2099
KU812 are basophils that were isolated from the peripheral blood of a 38-year-old, Asian, male patient with chronic myelogenous leukemia (cml). It can be used for immunology and immune system disorder research.