Learn More
Chemglass Life Sciences 500mL Kuderna-Danish Flask with Hooks
SDP
Catalog No. CG130803
Encompass_Preferred
500mL Kuderna-Danish Flask with Hooks. Lower body of flask has a hemispherical shape to afford continuous flow of heavier fractions to concentrator tube during reflux. Top joint is a 24/40 outer while the lower joint is a 19/22 inner.With hooks on the lower 19/22 inner joint.
Catalog No. CG130803 Supplier Chemglass Life Sciences Supplier No. CG130803
500mL Kuderna-Danish Flask with Hooks. Lower body of flask has a hemispherical shape to afford continuous flow of heavier fractions to concentrator tube during reflux. Top joint is a 24/40 outer while the lower joint is a 19/22 inner.With hooks on the lower 19/22 inner joint.