Learn More
Medchemexpress LLC HY-N4253 1mg Medchemexpress, Kudinoside D CAS:173792-61-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN42531MG
Encompass_Preferred
Medchemexpress, HY-N4253 1mg Kudinoside D CAS:173792-61-5 Kudinoside D is a main natural component of triterpenoid saponin derived from Ilex kudingcha. Kudinoside D suppresses adipogenesis through modulation of the AMPK pathway in 3T3-L1 adipocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2930