Learn More
Roboz Surgical KUEHNE FORCEPS 4IN ANGFLAT JAW
SDP
Catalog No. NC9588737
Encompass
Kuehne Forceps; Flat Jaw; Angled; 4 in. Length The Kuehne Micro Dissecting Forceps are recommended for picking up slides and cover glass. They have a unique thin flat, angled, bill-like tip that allows the fine glass to slip onto its tip easily. Th
Catalog No. NC9588737 Supplier Roboz Surgical Supplier No. RS8260
Kuehne Forceps; Flat Jaw; Angled; 4 in. Length The Kuehne Micro Dissecting Forceps are recommended for picking up slides and cover glass. They have a unique thin flat, angled, bill-like tip that allows the fine glass to slip onto its tip easily. Th