Learn More
Kuhner Shaker Inc KUHNER GAS DIVIDER 105519
Catalog No. NC2734324
Encompass
Kuhner GasDivider (105519)..- Gasing unit..- Flask..- 3m tubing (3.2/6.4mm)..- 3m tubing (1.6/4.8mm)..- Filter 0.4 micrometer..- Black connector set to Kuhner FlowCon..- Luerlock male..- 1m blue PUR tubing (4/6mm)
Catalog No. NC2734324 Supplier Kuhner Shaker Inc Supplier No. SMDG00001
Kuhner GasDivider (105519)..- Gasing unit..- Flask..- 3m tubing (3.2/6.4mm)..- 3m tubing (1.6/4.8mm)..- Filter 0.4 micrometer..- Black connector set to Kuhner FlowCon..- Luerlock male..- 1m blue PUR tubing (4/6mm)