Learn More
Kuhner Shaker Inc KUHNER KELVIN INCUBATOR
Catalog No. 502569938
Encompass
LS-Z Shaker + Kuhner Kelvin IncubatorInner chamber complete stainless steelHeating power : 400 WGas volume : 130 LAir circulation : 150 m3/hTray size E : 420x420mmUsable height: 400mmAmbient temp. : 10°C to 35°CTemp. range : ambient +5°C - 50°CTemp. accuracy : +/- 0.5°CWeight : 80.5 kgDimensions : 558x746x672mm w/o PowerCube: 558x613x672mmInterface : CAN Bus+ Optional : USB / EthernetEquipped with LED interior light andpolycarbonate doorIncludes:Kuhner Kelvin+Shaking UnitPower Cube
Catalog No. 50-256-9938 Supplier Kuhner Shaker Inc Supplier No. SMZ1270
Add to Cart
Add to Cart
Request a Quote
LS-Z Shaker + Kuhner Kelvin IncubatorInner chamber complete stainless steelHeating power : 400 WGas volume : 130 LAir circulation : 150 m3/hTray size E : 420x420mmUsable height: 400mmAmbient temp. : 10°C to 35°CTemp. range : ambient +5°C - 50°CTemp. accuracy : +/- 0.5°CWeight : 80.5 kgDimensions : 558x746x672mm w/o PowerCube: 558x613x672mmInterface : CAN Bus+ Optional : USB / EthernetEquipped with LED interior light andpolycarbonate doorIncludes:Kuhner Kelvin+Shaking UnitPower Cube