Learn More
Medchemexpress LLC Kukoamine B | 164991-67-7 | 5 MG
SDP
Catalog No. 5000420687
Encompass_Preferred
Kukoamine B is a spermine alkaloid and a potent dual LPS and CpG DNA inhibitor. It exhibits anti-inflammatory, anti-diabetic, anti-oxidant, anti-osteoporotic, and neuroprotective effects, and is being studied for its potential in sepsis treatment.
  • Increases cell viability and prevents plasma membrane damage in SH-SY5Y cells.
  • Attenuates H2O2-induced apoptosis and mitochondrial membrane potential (MMP) loss.
  • Demonstrates anti-oxidative stress effects by modulating MAPKs and PI3K-AKT signaling pathways.
  • Increases osteoblast differentiation and mineralized nodule formation.
  • Dual inhibitor of LPS- and CpG DNA-induced TNF-α and IL-6 release.
  • Reduces inflammation and blood glucose.
  • Exhibits anti-osteoporotic effects.
  • Protects against E. coli challenges and reduces circulatory levels of LPS and TNF-α.
Catalog No. 50-004-20687 Supplier Medchemexpress LLC Supplier No. HYN23935MG
Add to Cart
Add to Cart
Kukoamine B is a spermine alkaloid and a potent dual LPS and CpG DNA inhibitor. It exhibits anti-inflammatory, anti-diabetic, anti-oxidant, anti-osteoporotic, and neuroprotective effects, and is being studied for its potential in sepsis treatment.
  • Increases cell viability and prevents plasma membrane damage in SH-SY5Y cells.
  • Attenuates H2O2-induced apoptosis and mitochondrial membrane potential (MMP) loss.
  • Demonstrates anti-oxidative stress effects by modulating MAPKs and PI3K-AKT signaling pathways.
  • Increases osteoblast differentiation and mineralized nodule formation.
  • Dual inhibitor of LPS- and CpG DNA-induced TNF-α and IL-6 release.
  • Reduces inflammation and blood glucose.
  • Exhibits anti-osteoporotic effects.
  • Protects against E. coli challenges and reduces circulatory levels of LPS and TNF-α.