Learn More
Medchemexpress LLC Kukoamine B | 164991-67-7 | MFCD29904539 | 99.9% | 530.66 g/mol | C28H42N4O6 | 10 MG
SDP
Catalog No. 5000288922
Encompass_Preferred
Kukoamine B is a spermine alkaloid supplied as a high-purity research chemical that acts as a dual inhibitor of lipopolysaccharide (LPS) and CpG DNA for biochemical and cellular studies.
  • Dual inhibitor of LPS and CpG DNA.
  • High purity (~99.9%) suitable for analytical and biological assays.
  • Available in small research pack sizes, including 10 MG powder and 10 mM solutions.
  • Molecular formula C28H42N4O6; molecular weight 530.66 g/mol.
  • Used in anti-inflammatory, antioxidant, and metabolic research applications.
Catalog No. 50-002-88922 Supplier Medchemexpress LLC Supplier No. HYN239310MG
Kukoamine B is a spermine alkaloid supplied as a high-purity research chemical that acts as a dual inhibitor of lipopolysaccharide (LPS) and CpG DNA for biochemical and cellular studies.
  • Dual inhibitor of LPS and CpG DNA.
  • High purity (~99.9%) suitable for analytical and biological assays.
  • Available in small research pack sizes, including 10 MG powder and 10 mM solutions.
  • Molecular formula C28H42N4O6; molecular weight 530.66 g/mol.
  • Used in anti-inflammatory, antioxidant, and metabolic research applications.