Learn More
Medchemexpress LLC HY-N2279 5mg Medchemexpress, Kurarinone CAS:34981-26-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22795MG
Encompass_Preferred
Medchemexpress, HY-N2279 5mg Kurarinone CAS:34981-26-5 Kurarinone, a flavanoid derived from shrub Sophora flavescens, inhibits the process of experimental autoimmune encephalomyelitis via blocking Th1 and Th17 cell differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0380