Learn More
Medchemexpress LLC HY-N2278 5mg Medchemexpress, Kushenol A CAS:99217-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22785MG
Encompass_Preferred
Medchemexpress, HY-N2278 5mg Kushenol A CAS:99217-63-7 Kushenol A is isolated from the root of Sophora flavescent, a non-competitive tyrosinase inhibitor with IC50 and Ki values of 1.1 μM and 0.4 μM, respectively.Kushenol A is a flavonoid antioxidant, has inhibitory effects on alpha-glucosidase and beta-amylase.Kushenol A is confirmed as potential inhibitors of enzymes targeted by cosmetics for skin whitening and aging. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0379