Learn More
Medchemexpress LLC Kushenol A | 99217-63-7 | 1 MG
SDP
Catalog No. 5000420451
Encompass_Preferred
Kushenol A is a compound isolated from the root of Sophora flavescent. It is classified as a flavonoid antioxidant.
  • Tyrosinase inhibitor
  • Glycosidase inhibitor
  • Antioxidant activity
  • Cytotoxic effects against NSCLC cells (A549 and NCI-H226 cell lines)
  • High purity (99.98%)
Catalog No. 50-004-20451 Supplier Medchemexpress LLC Supplier No. HYN22781MG
Add to Cart
Add to Cart
Kushenol A is a compound isolated from the root of Sophora flavescent. It is classified as a flavonoid antioxidant.
  • Tyrosinase inhibitor
  • Glycosidase inhibitor
  • Antioxidant activity
  • Cytotoxic effects against NSCLC cells (A549 and NCI-H226 cell lines)
  • High purity (99.98%)