Learn More
Medchemexpress LLC Kushenol A | 99217-63-7 | 1 ML
SDP
Catalog No. 5000421714
Encompass_Preferred
Kushenol A (Leachianone E) is a flavonoid antioxidant isolated from the root of Sophora flavescent. This compound acts as a non-competitive tyrosinase inhibitor, blocking the conversion of L-tyrosine to L-DOPA. It also shows inhibitory effects on alpha-glucosidase and β-amylase, making it a potential candidate for cosmetic applications in skin whitening and anti-aging.
  • Isolated from the root of Sophora flavescent
  • Non-competitive tyrosinase inhibitor
  • Blocks conversion of L-tyrosine to L-DOPA
  • Flavonoid antioxidant
  • Inhibits alpha-glucosidase and β-amylase
  • Potential for skin whitening and anti-aging applications
  • Highly potent inhibitory activity on ABTS•+ radical scavenging
  • Exhibits cytotoxic effects against NSCLC cells
Catalog No. 50-004-21714 Supplier Medchemexpress LLC Supplier No. HYN227810MM/1ML
Kushenol A (Leachianone E) is a flavonoid antioxidant isolated from the root of Sophora flavescent. This compound acts as a non-competitive tyrosinase inhibitor, blocking the conversion of L-tyrosine to L-DOPA. It also shows inhibitory effects on alpha-glucosidase and β-amylase, making it a potential candidate for cosmetic applications in skin whitening and anti-aging.
  • Isolated from the root of Sophora flavescent
  • Non-competitive tyrosinase inhibitor
  • Blocks conversion of L-tyrosine to L-DOPA
  • Flavonoid antioxidant
  • Inhibits alpha-glucosidase and β-amylase
  • Potential for skin whitening and anti-aging applications
  • Highly potent inhibitory activity on ABTS•+ radical scavenging
  • Exhibits cytotoxic effects against NSCLC cells