Learn More
MSC Claw Cover Cut-Resistant Glove, Small, ANSI A7, 1 ea
Catalog No. 5000686881
Encompass
These medium-weight, cut-resistant gloves provide superior hand protection in environments where sharp tools are present. Featuring a durable seamless knit construction, these gloves incorporate stainless steel, silica fiber, and Dyneema® for maximum resistance to cuts and slashes while maintaining flexibility and breathability. Designed for versatility, they are suitable for food service, processing, and metal handling applications.
  • Seamless knit construction for improved comfort, fit, and dexterity
  • Dyneema® blend offers enhanced performance without sacrificing flexibility
  • Ambidextrous design allows for use on either hand to extend wear
  • Non-latex cuff resists blowout during frequent laundering
  • Antimicrobial material is non-toxic and food-contact compliant
  • Shrink-resistant and launderable for repeated use
Catalog No. 50-006-86881 Supplier MSC Supplier No. 08437477
These medium-weight, cut-resistant gloves provide superior hand protection in environments where sharp tools are present. Featuring a durable seamless knit construction, these gloves incorporate stainless steel, silica fiber, and Dyneema® for maximum resistance to cuts and slashes while maintaining flexibility and breathability. Designed for versatility, they are suitable for food service, processing, and metal handling applications.
  • Seamless knit construction for improved comfort, fit, and dexterity
  • Dyneema® blend offers enhanced performance without sacrificing flexibility
  • Ambidextrous design allows for use on either hand to extend wear
  • Non-latex cuff resists blowout during frequent laundering
  • Antimicrobial material is non-toxic and food-contact compliant
  • Shrink-resistant and launderable for repeated use