Learn More
Enterprise Technology Solutions Kutol Products Hand Sanitizer, No Alcohol, 1000 mL, 6/CT, Clear
SDP
Catalog No. 5000810995
Encompass_Preferred
Health Guard Foaming No Alcohol Hand Sanitizer is an effective, convenient way to sanitize hands when soap and water are not available. Sanitizer is formulated with 0.13 percent benzalkonium chloride to eliminate 99.99 percent of common germs within 15 seconds. It does not dry out skin like alcohol sanitizers can and is dye-free. Hand sanitizer is perfect for use where a non-alcohol product is desired including schools, public facilities, stadiums and arenas and nursing homes. Sanitizer is designed for the EZ Foam Wall Mount (sold separately).
Catalog No. 50-008-10995 Supplier Enterprise Technology Solutions Supplier No. KUT68241
Health Guard Foaming No Alcohol Hand Sanitizer is an effective, convenient way to sanitize hands when soap and water are not available. Sanitizer is formulated with 0.13 percent benzalkonium chloride to eliminate 99.99 percent of common germs within 15 seconds. It does not dry out skin like alcohol sanitizers can and is dye-free. Hand sanitizer is perfect for use where a non-alcohol product is desired including schools, public facilities, stadiums and arenas and nursing homes. Sanitizer is designed for the EZ Foam Wall Mount (sold separately).