Learn More
Enterprise Technology Solutions Kutol Products Hand Soap, Antibacterial, 1000 mL, 6/CT, Amber
SDP
Catalog No. 5000828910
Encompass_Preferred
Health Guard Foaming Advanced Antibacterial Hand Soap contains 0.13 percent benzalkonium chloride to kill 99.99 percent of the most common germs that can cause illness. This rich foam cleans away light to medium soils. It's specially formulated without triclosan and contains aloe and Vitamin E to help nourish the skin. Soap is designed for the EZ Foam Wall Mount (sold separately).
Catalog No. 50-008-28910 Supplier Enterprise Technology Solutions Supplier No. KUT21341
Health Guard Foaming Advanced Antibacterial Hand Soap contains 0.13 percent benzalkonium chloride to kill 99.99 percent of the most common germs that can cause illness. This rich foam cleans away light to medium soils. It's specially formulated without triclosan and contains aloe and Vitamin E to help nourish the skin. Soap is designed for the EZ Foam Wall Mount (sold separately).