Learn More
Kuhner Shaker Inc KUUHNER AERATION CAP 500 ML 1
Catalog No. 502570111
Encompass
Kuuhner Aeration Cap 500 ml (105499)..1x AerationCap for 500mL Corning flask..2x Luerlock..1x Sealing ring
Catalog No. 50-257-0111 Supplier Kuhner Shaker Inc Supplier No. SMDG00015
Kuuhner Aeration Cap 500 ml (105499)..1x AerationCap for 500mL Corning flask..2x Luerlock..1x Sealing ring