Learn More
Medchemexpress LLC HY-N2300 2mg Medchemexpress, Kuwanon A CAS:62949-77-3 Purity:>98%
SDP
Catalog No. 501933904
Encompass_Preferred
Medchemexpress, HY-N2300 2mg Kuwanon A CAS:62949-77-3 Kuwanon A is a flavone derivative isolated from the root barks of the mulberry tree (Morus alba L.); inhibits nitric oxide production with an IC 50 of 10.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3904 Supplier Medchemexpress LLC Supplier No. HYN23002MG
Medchemexpress, HY-N2300 2mg Kuwanon A CAS:62949-77-3 Kuwanon A is a flavone derivative isolated from the root barks of the mulberry tree (Morus alba L.); inhibits nitric oxide production with an IC 50 of 10.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.