Learn More
Medchemexpress LLC Kuwanon E | 68401-05-8 | 1 MG
SDP
Catalog No. 5000417898
Encompass_Preferred
Kuwanon E is a flavonoid isolated from *Morus alba*. It exhibits cytotoxicity to human monocytic leukemic cell lines and reduces the level of IL-1β. This compound demonstrates cytotoxic activity against the THP-1 human monocytic leukemic cell line with an IC50 of 4.0 ± 0.08 μM.
  • Isolated from *Morus alba*
  • Cytotoxic to human monocytic leukemic cell lines
  • Reduces the level of IL-1β
  • Inhibits SERCA1 in New Zealand rabbit skeletal muscle SR vesicles
  • For research use only
Catalog No. 50-004-17898 Supplier Medchemexpress LLC Supplier No. HYN35141MG
Kuwanon E is a flavonoid isolated from *Morus alba*. It exhibits cytotoxicity to human monocytic leukemic cell lines and reduces the level of IL-1β. This compound demonstrates cytotoxic activity against the THP-1 human monocytic leukemic cell line with an IC50 of 4.0 ± 0.08 μM.
  • Isolated from *Morus alba*
  • Cytotoxic to human monocytic leukemic cell lines
  • Reduces the level of IL-1β
  • Inhibits SERCA1 in New Zealand rabbit skeletal muscle SR vesicles
  • For research use only