Learn More
Medchemexpress LLC Kuwanon E | 68401-05-8 | 5 MG
SDP
Catalog No. 5000421057
Encompass_Preferred
Kuwanon E is a flavonoid isolated from Morus alba.
  • Cytotoxic to human monocytic leukemic cell lines.
  • Reduces the level of IL-1β.
  • Shows cytotoxic activity against THP-1 human monocytic leukemic cell line, with an IC50 of 4.0±0.08 μM.
  • Inhibits SERCA1 in New Zealand rabbit skeletal muscle SR vesicles.
Catalog No. 50-004-21057 Supplier Medchemexpress LLC Supplier No. HYN35145MG
Kuwanon E is a flavonoid isolated from Morus alba.
  • Cytotoxic to human monocytic leukemic cell lines.
  • Reduces the level of IL-1β.
  • Shows cytotoxic activity against THP-1 human monocytic leukemic cell line, with an IC50 of 4.0±0.08 μM.
  • Inhibits SERCA1 in New Zealand rabbit skeletal muscle SR vesicles.