Learn More
Medchemexpress LLC Kuwanon G | 75629-19-5 | 5 MG
SDP
Catalog No. 5000418777
Encompass_Preferred
Kuwanon G is a flavonoid compound and a bombesin receptor antagonist with various biological activities. It demonstrates strong antibacterial, anti-tumor, anti-inflammatory, antioxidant, anti-atherosclerotic, and neuroprotective effects, making it suitable for research on various diseases such as gastric cancer and atherosclerosis.
  • Strong antibacterial activity against oral pathogens.
  • Induces apoptosis and inhibits proliferation, migration, and invasion of tumor cells.
  • Inhibits mRNA levels of inflammatory factors.
  • Protects cells from advanced glycation end product-induced damage.
  • Attenuates atherosclerosis by regulating LXRα-ABCA1/ABCG1 and inhibiting NFκB activity.
  • Demonstrates neuroprotective effects.
  • Acts as a non-peptide bombesin receptor antagonist.
Catalog No. 50-004-18777 Supplier Medchemexpress LLC Supplier No. HYN42475MG
Kuwanon G is a flavonoid compound and a bombesin receptor antagonist with various biological activities. It demonstrates strong antibacterial, anti-tumor, anti-inflammatory, antioxidant, anti-atherosclerotic, and neuroprotective effects, making it suitable for research on various diseases such as gastric cancer and atherosclerosis.
  • Strong antibacterial activity against oral pathogens.
  • Induces apoptosis and inhibits proliferation, migration, and invasion of tumor cells.
  • Inhibits mRNA levels of inflammatory factors.
  • Protects cells from advanced glycation end product-induced damage.
  • Attenuates atherosclerosis by regulating LXRα-ABCA1/ABCG1 and inhibiting NFκB activity.
  • Demonstrates neuroprotective effects.
  • Acts as a non-peptide bombesin receptor antagonist.