Learn More
Medchemexpress LLC KV2 channel inhibitor-1 | 689297-68-5 | 98.7% | 396.89 | 5 MG
SDP
Catalog No. 5000404783
Encompass_Preferred
KV2 channel inhibitor-1 is a selective inhibitor for KV2 channels, with IC50 values of 0.2 μM for KV2.1 and 0.41 μM for KV2.2. It shows good selectivity, being >10-fold selective over NaV channels and other KV channels, and exhibiting weak activity on CaV channels. It also possesses good selectivity over KV1.2 (IC50>10 μM). In vitro, KV2 channel inhibitor-1 (compound A1) inhibits the KV current in rat insulinoma INS-1 cells, blocking the INS-1 KV current by an average of 71% at 0.3 μM and 84% at 3 μM.
  • Selective KV2 channel inhibitor.
  • IC50s: 0.2 μM (KV2.1), 0.41 μM (KV2.2).
  • Good selectivity over KV1.2 (IC50>10 μM).
  • >10-fold selective over NaV channels and other KV channels.
  • Displays weak activity on CaV channels.
Catalog No. 50-004-04783 Supplier Medchemexpress LLC Supplier No. HY441535MG
KV2 channel inhibitor-1 is a selective inhibitor for KV2 channels, with IC50 values of 0.2 μM for KV2.1 and 0.41 μM for KV2.2. It shows good selectivity, being >10-fold selective over NaV channels and other KV channels, and exhibiting weak activity on CaV channels. It also possesses good selectivity over KV1.2 (IC50>10 μM). In vitro, KV2 channel inhibitor-1 (compound A1) inhibits the KV current in rat insulinoma INS-1 cells, blocking the INS-1 KV current by an average of 71% at 0.3 μM and 84% at 3 μM.
  • Selective KV2 channel inhibitor.
  • IC50s: 0.2 μM (KV2.1), 0.41 μM (KV2.2).
  • Good selectivity over KV1.2 (IC50>10 μM).
  • >10-fold selective over NaV channels and other KV channels.
  • Displays weak activity on CaV channels.