Learn More
Medchemexpress LLC Kv2 channel inhibitor-1 | 689297-68-5 | MFCD32631937 | 98.7% | 396.89 | C20H17ClN4OS | 10 MG
SDP
Catalog No. 5000298549
Encompass_Preferred
KV2 channel inhibitor-1 is a selective small-molecule inhibitor of KV2 family potassium channels, with potent activity against KV2.1 (IC50 0.2 μM) and KV2.2 (IC50 0.41 μM). It is supplied as a solid research compound for in vitro electrophysiology and pharmacological profiling, with reported selectivity data versus other voltage-gated ion channels.
  • Selective inhibition of KV2.1 and KV2.2 (IC50 0.2 μM and 0.41 μM).
  • High purity suitable for research use.
  • Provided as a white to off-white solid for assay work.
  • Includes selectivity data against KV, CaV, and NaV channels.
  • Available in small milligram quantities for in vitro studies.
Catalog No. 50-002-98549 Supplier Medchemexpress LLC Supplier No. HY4415310MG
KV2 channel inhibitor-1 is a selective small-molecule inhibitor of KV2 family potassium channels, with potent activity against KV2.1 (IC50 0.2 μM) and KV2.2 (IC50 0.41 μM). It is supplied as a solid research compound for in vitro electrophysiology and pharmacological profiling, with reported selectivity data versus other voltage-gated ion channels.
  • Selective inhibition of KV2.1 and KV2.2 (IC50 0.2 μM and 0.41 μM).
  • High purity suitable for research use.
  • Provided as a white to off-white solid for assay work.
  • Includes selectivity data against KV, CaV, and NaV channels.
  • Available in small milligram quantities for in vitro studies.