Learn More
TARGETMOL CHEMICALS INC Kv3 modulator 1 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T117871MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Kv3 modulator 1 is a voltage-gated potassium channel Kv3 modulator that can be used to study neurologic-level diseases. Purity 99.1%
Catalog No. 50-287-9642