Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kv3 modulator 2 | 2101321-76-8 | 99.9% | 25 MG
SDP

Catalog No. 5000342390
Encompass_Preferred

Kv3 modulator 2 is a potent Kv3 channels modulator, extracted from patent WO2018109484A1. It exhibits analgesic activity and is utilized in the prophylaxis or treatment of related disorders. This compound is for research use only and is not intended for patient use. It has a molecular weight of 379.41 and a chemical formula of C21H21N3O4, appearing as a solid that is white to off-white in color.

  • Potent Kv3 channels modulator
  • Exhibits analgesic activity
  • Used in the prophylaxis or treatment of related disorders
  • For research use only
  • Molecular weight: 379.41
  • Chemical formula: C21H21N3O4
  • Appearance: Solid, white to off-white
  • Shipped at room temperature in the continental US
  • Storage conditions: 4°C sealed, away from moisture and light; in solvent: -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light)
  • Solubility: 100 mg/mL in DMSO

Catalog No. 50-003-42390 Supplier Medchemexpress LLC Supplier No. 25MGHY128829
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kv3 modulator 2 is a potent Kv3 channels modulator, extracted from patent WO2018109484A1. It exhibits analgesic activity and is utilized in the prophylaxis or treatment of related disorders. This compound is for research use only and is not intended for patient use. It has a molecular weight of 379.41 and a chemical formula of C21H21N3O4, appearing as a solid that is white to off-white in color.

  • Potent Kv3 channels modulator
  • Exhibits analgesic activity
  • Used in the prophylaxis or treatment of related disorders
  • For research use only
  • Molecular weight: 379.41
  • Chemical formula: C21H21N3O4
  • Appearance: Solid, white to off-white
  • Shipped at room temperature in the continental US
  • Storage conditions: 4°C sealed, away from moisture and light; in solvent: -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light)
  • Solubility: 100 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.