Learn More
Medchemexpress LLC Kv3 modulator 2 | 2101321-76-8 | 99.9% | 379.41 g·mol⁻¹ | C21H21N3O4 | 5 MG
SDP
Catalog No. 5000264259
Encompass_Preferred
Kv3 modulator 2 is a small-molecule modulator of Kv3 voltage-gated potassium channels with reported analgesic activity. It is provided for laboratory research use and is characterized by a defined molecular formula and molecular weight, and high purity suitable for preclinical studies.
  • High purity (99.9%).
  • Available in small research quantities (example: 5 mg).
  • Molecular formula C21H21N3O4; molecular weight 379.41 g·mol⁻¹.
  • Reported Kv3 channel modulatory activity with analgesic effects.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-64259 Supplier Medchemexpress LLC Supplier No. HY1288295MG
Kv3 modulator 2 is a small-molecule modulator of Kv3 voltage-gated potassium channels with reported analgesic activity. It is provided for laboratory research use and is characterized by a defined molecular formula and molecular weight, and high purity suitable for preclinical studies.
  • High purity (99.9%).
  • Available in small research quantities (example: 5 mg).
  • Molecular formula C21H21N3O4; molecular weight 379.41 g·mol⁻¹.
  • Reported Kv3 channel modulatory activity with analgesic effects.
  • For research use only; not for human or clinical use.