Learn More
Medchemexpress LLC KVA-D-88 | 2410550-31-9 | 99.8% | 382.19 | 1 ML
SDP
Catalog No. 5000350753
Encompass_Preferred
KVA-D-88 is a blood-brain barrier-permeable PDE4 inhibitor with IC50 values of 140 and 880 nM for PDE4B and PDE4D, respectively. It inhibits cocaine-induced hyperlocomotor activity and sensitization, selectively inhibiting PDE4B and reducing cAMP degradation, thereby modulating neuroadaptive responses. It can be used in the study of cocaine addiction.
  • Blood-brain barrier-permeable PDE4 inhibitor
  • Inhibits cocaine-induced hyperlocomotor activity and sensitization
  • Selectively inhibits PDE4B and reduces cAMP degradation
  • Modulates neuroadaptive responses
  • Useful in the study of cocaine addiction
Catalog No. 50-003-50753 Supplier Medchemexpress LLC Supplier No. HY13917810MM/1ML
KVA-D-88 is a blood-brain barrier-permeable PDE4 inhibitor with IC50 values of 140 and 880 nM for PDE4B and PDE4D, respectively. It inhibits cocaine-induced hyperlocomotor activity and sensitization, selectively inhibiting PDE4B and reducing cAMP degradation, thereby modulating neuroadaptive responses. It can be used in the study of cocaine addiction.
  • Blood-brain barrier-permeable PDE4 inhibitor
  • Inhibits cocaine-induced hyperlocomotor activity and sensitization
  • Selectively inhibits PDE4B and reduces cAMP degradation
  • Modulates neuroadaptive responses
  • Useful in the study of cocaine addiction