Learn More
Medchemexpress LLC KVS0001 | 3056394-59-0 | 99.1% | 579.07 | 50 MG
SDP
Catalog No. 5000393073
Encompass_Preferred
KVS0001 is a selective SMG1 inhibitor that elevates the expression of transcripts and proteins resulting from truncating mutations. It increases the presentation of immune-targetable HLA class I-associated peptides from nonsense-mediated decay (NMD)-downregulated proteins on the surface of cancer cells. This compound exerts anti-tumor properties and can be studied in research for NMD-related diseases, including cancer and inherited diseases.
  • Inhibits SMG1 protein more selectively than other kinases.
  • Subverts nonsense-mediated decay (NMD)-mediated downregulation of mutant transcripts and proteins.
  • Increases interferon-gamma (IFN-γ) release and killing of target cancer cells.
  • Significantly increases RNA in genes with truncating mutations.
  • Slows tumor growth in C57BL/6N and BALB/c mouse models.
  • Increases transcript levels in genes with truncating mutations in xenograft tumors.
Catalog No. 50-003-93073 Supplier Medchemexpress LLC Supplier No. HY16111150MG
KVS0001 is a selective SMG1 inhibitor that elevates the expression of transcripts and proteins resulting from truncating mutations. It increases the presentation of immune-targetable HLA class I-associated peptides from nonsense-mediated decay (NMD)-downregulated proteins on the surface of cancer cells. This compound exerts anti-tumor properties and can be studied in research for NMD-related diseases, including cancer and inherited diseases.
  • Inhibits SMG1 protein more selectively than other kinases.
  • Subverts nonsense-mediated decay (NMD)-mediated downregulation of mutant transcripts and proteins.
  • Increases interferon-gamma (IFN-γ) release and killing of target cancer cells.
  • Significantly increases RNA in genes with truncating mutations.
  • Slows tumor growth in C57BL/6N and BALB/c mouse models.
  • Increases transcript levels in genes with truncating mutations in xenograft tumors.