Learn More
Apexbio Technology LLC KW 2449 1000669-72-6 10mg
SDP
Catalog No. 5000096257
Encompass_Preferred
KW 2449 (CAS 1000669-72-6) is a multi-targeted kinase inhibitor with potent activity against FLT3 (IC50 6 6 nM) ABL (14 nM) Aurora kinases (4 nM) and FGFR (48 nM) It inhibits both wild-type and T315I-mutated ABL tyrosine kinase In vitro KW 2449 disrupts proliferation through modulation of distinct signaling pathways it downregulates FLT3/STAT5 phosphorylation and induces G1-phase arrest and apoptosis in FLT3-mutated leukemia cells while promoting G2/M arrest via reduced histone H3 phosphorylation in FLT3 wild-type cells KW 2449 is applied in research investigating kinase-driven leukemias and resistance mechanisms to targeted therapies
Catalog No. 50-000-96257 Supplier Apexbio Technology LLC Supplier No. A412310
KW 2449 (CAS 1000669-72-6) is a multi-targeted kinase inhibitor with potent activity against FLT3 (IC50 6 6 nM) ABL (14 nM) Aurora kinases (4 nM) and FGFR (48 nM) It inhibits both wild-type and T315I-mutated ABL tyrosine kinase In vitro KW 2449 disrupts proliferation through modulation of distinct signaling pathways it downregulates FLT3/STAT5 phosphorylation and induces G1-phase arrest and apoptosis in FLT3-mutated leukemia cells while promoting G2/M arrest via reduced histone H3 phosphorylation in FLT3 wild-type cells KW 2449 is applied in research investigating kinase-driven leukemias and resistance mechanisms to targeted therapies