Learn More
Medchemexpress LLC KW-2449 | 1000669-72-6 | 99.9% | 332.40 | 10 MG
SDP
Catalog No. 5000364834
Encompass_Preferred
KW-2449 is a multi-targeted kinase inhibitor that targets FLT3, ABL, ABLT315I, and Aurora kinase with IC50 values of 6.6, 14, 4, and 48 nM, respectively. It is intended for research use only.
  • Targets FLT3, ABL, ABLT315I, and Aurora kinase.
  • Exhibits antiproliferative activity against imatinib-resistant K562 cells.
  • Inhibits FLT3-mediated STAT5 phosphorylation.
  • Increases G1 phase cells and promotes apoptosis.
  • Suppresses tumor growth in FLT3-mutated xenograft models.
  • Helps release imatinib resistance.
Catalog No. 50-003-64834 Supplier Medchemexpress LLC Supplier No. 10MGHY10339
KW-2449 is a multi-targeted kinase inhibitor that targets FLT3, ABL, ABLT315I, and Aurora kinase with IC50 values of 6.6, 14, 4, and 48 nM, respectively. It is intended for research use only.
  • Targets FLT3, ABL, ABLT315I, and Aurora kinase.
  • Exhibits antiproliferative activity against imatinib-resistant K562 cells.
  • Inhibits FLT3-mediated STAT5 phosphorylation.
  • Increases G1 phase cells and promotes apoptosis.
  • Suppresses tumor growth in FLT3-mutated xenograft models.
  • Helps release imatinib resistance.