Learn More
TARGETMOL CHEMICALS INC KW-2449 25MG
SDP
Catalog No. 502580873
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KW-2449 is a multiple-targeted inhibitor mostly for Flt3 (IC50 6.6 nM) modestly effective to Bcr-Abl FGFR1 and Aurora A; little inhibitory on PDGFR(beta) IGF-1R EGFR. purity: 99%
Catalog No. 50-258-0873 Supplier TARGETMOL CHEMICALS INC Supplier No. T234125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KW-2449 is a multiple-targeted inhibitor mostly for Flt3 (IC50 6.6 nM) modestly effective to Bcr-Abl FGFR1 and Aurora A; little inhibitory on PDGFR(beta) IGF-1R EGFR. purity: 99%