Learn More
Selleck Chemical LLC KW-2478 S2685-5mg
SDP
Catalog No. 503112504
Encompass_Preferred
KW-2478 is a nonansamycin HSP90 inhibitor with IC50 of 3 8 nM Phase 1
Catalog No. 50-311-2504 Supplier Selleck Chemical LLC Supplier No. S26855MG
KW-2478 is a nonansamycin HSP90 inhibitor with IC50 of 3 8 nM Phase 1