Learn More
TARGETMOL CHEMICALS INC KW-2478 5MG
SDP
Catalog No. 502576492
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KW-2478 is a non-ansamycin HSP90 inhibitor with IC50 of 3.8 nM. purity: 98%
Catalog No. 50-257-6492 Supplier TARGETMOL CHEMICALS INC Supplier No. T65585MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KW-2478 is a non-ansamycin HSP90 inhibitor with IC50 of 3.8 nM. purity: 98%