Learn More
Medchemexpress LLC KW-8232 | 217813-15-5 | 98.1% | 705.26 | C37H41ClN4O6S | 5 MG
SDP
Catalog No. 5000214047
Encompass_Preferred
KW-8232 is an orally active anti-osteoporotic small molecule that reduces prostaglandin E2 (PGE2) biosynthesis and decreases markers of bone resorption. In animal models it increases femoral bone mineral density and lowers urinary calcium and collagen crosslinks. Supplied as a solid or as a pre-made DMSO solution for in vitro use.
  • Orally active anti-osteoporotic agent with PGE2 inhibition.
  • Reduces bone resorption markers such as pyridinoline and deoxypyridinoline.
  • Demonstrated to increase femoral bone mineral density in animal models.
  • High purity (98.1%) suitable for research applications.
  • Soluble in DMSO (approx. 125 mg/mL) for in vitro testing.
  • Available as solid and as prepared DMSO solution for convenience.
Catalog No. 50-002-14047 Supplier Medchemexpress LLC Supplier No. HY100304A5MG
KW-8232 is an orally active anti-osteoporotic small molecule that reduces prostaglandin E2 (PGE2) biosynthesis and decreases markers of bone resorption. In animal models it increases femoral bone mineral density and lowers urinary calcium and collagen crosslinks. Supplied as a solid or as a pre-made DMSO solution for in vitro use.
  • Orally active anti-osteoporotic agent with PGE2 inhibition.
  • Reduces bone resorption markers such as pyridinoline and deoxypyridinoline.
  • Demonstrated to increase femoral bone mineral density in animal models.
  • High purity (98.1%) suitable for research applications.
  • Soluble in DMSO (approx. 125 mg/mL) for in vitro testing.
  • Available as solid and as prepared DMSO solution for convenience.